Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-tetrahydrofuran complex, 1M soln. in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog Number AA43291AP
Click to view available options
Quantity:
100 mL
500 mL
14044-65-6
C4H11BO
85.94
MFCD00012429
RMCYTHFAWCWRFA-UHFFFAOYSA-N
borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf
11062302
boron;oxolane
B.C1CCOC1
14044-65-6
C4H11BO
85.94
MFCD00012429
RMCYTHFAWCWRFA-UHFFFAOYSA-N
borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf
11062302
boron;oxolane
B.C1CCOC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.878 g/mL
Flash Point -21°C (-8°F)
Packaging ChemSeal™ bottles
Linear Formula BH3/(CH2)4O
Quantity 500 mL
UN Number UN3148
Beilstein 3668402
Sensitivity Air and moisture sensitive
Solubility Information Miscible and reacts violently with water.
Formula Weight 85.94
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Physical Form Liquid
Chemical Name or Material Borane-tetrahydrofuran complex
Show More Show Less
Hazard Category H225-H260-H302-H315-H318-H335-H336-H351
Hazard Statement GHS H Statement
H225-H260-H318-H351-H302-H335-H315
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P335+P334
DOTInformation Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.
EINECSNumber 237-881-8
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Argon
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit