Alfa Aesar™ Borane-tetrahydrofuran complex, 1M soln. in THF, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 43291AE

Quantity 100mL

 View more versions of this product

Catalog No. AA43291AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14044-65-6
    Linear Formula BH3/(CH2)4O
    Formula Weight 85.94
    MDL Number MFCD00012429
    UN Number UN3148
    Beilstein 3668402
    Synonym BTHF
    Chemical Name or Material Borane-tetrahydrofuran complex

    Specifications

    Formula Weight 85.94
    Health Hazard 2 GHS H Statement
    H225-H260-H318-H351-H302-H335-H315
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes serious eye damage.
    Suspected of causing cancer.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00012429
    UN Number UN3148
    Density 0.878g/mL
    Packaging ChemSeal™ bottles
    Color Colorless
    Synonym BTHF
    Quantity 100mL
    Sensitivity Air & Moisture Sensitive
    Health Hazard 1 Danger
    Storage Note 1 2° to 8°C
    DOT Information Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.
    TSCA TSCA
    Physical Form Liquid
    Linear Formula BH3/(CH2)4O
    CAS 14044-65-6
    Beilstein 3668402
    Concentration 1M soln. in THF
    Chemical Name or Material Borane-tetrahydrofuran complex

    GHS H Statement
    H225-H260-H318-H351-H302-H335-H315
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes serious eye damage.
    Suspected of causing cancer.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    2° to 8°C

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit