Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Borane-triethylamine complex, 96%, packaged under Argon in resealable ChemSeal™ bottles

As a reducing agent and in hydroboration reactions

Manufacturer:  Alfa Aesar™ 4296214

Quantity 25g

 View more versions of this product

Catalog No. AA4296214

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1722-26-5
(C2H5)3N·BH3
MFCD00012423
Borane-triethylamine complex
C6H15N·BH3
115.03
UN2924

Specifications

1722-26-5
ChemSeal™ bottles
C6H15N·BH3
115.03
UN2924
Colorless to pale yellow
25g
Fish-like
Borane-triethylamine complex
0.777g/mL
Liquid
(C2H5)3N·BH3
MFCD00012423
−4°C
97°C (12mmHg)
−6°C (21°F)
Moisture Sensitive
96%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit