Alfa Aesar™ Borane-triethylamine complex, 96%, packaged under Argon in resealable ChemSeal™ bottles

As a reducing agent and in hydroboration reactions

Manufacturer: Alfa Aesar™ 4296222

Quantity 100g

 View more versions of this product

Catalog No. AA4296222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1722-26-5
    Molecular Formula C6H15N·BH3
    Linear Formula (C2H5)3N·BH3
    Formula Weight 115.03
    MDL Number MFCD00012423
    UN Number UN2924
    Chemical Name or Material Borane-triethylamine complex

    Specifications

    Formula Weight 115.03
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 1722-26-5
    Physical Form Liquid
    Linear Formula (C2H5)3N·BH3
    Boiling Point 97°C (12mmHg)
    Odor Fish-like
    Health Hazard 1 Danger
    Molecular Formula C6H15N·BH3
    Melting Point −4°C
    Packaging ChemSeal™ bottles
    TSCA TSCA
    MDL Number MFCD00012423
    UN Number UN2924
    Quantity 100g
    Density 0.777g/mL
    Health Hazard 2 GHS H Statement
    H225-H314-H318-H302-H312-H332
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Color Colorless to pale yellow
    Flash Point −6°C (21°F)
    Sensitivity Moisture Sensitive
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Chemical Name or Material Borane-triethylamine complex
    Assay 96%

    GHS H Statement
    H225-H314-H318-H302-H312-H332
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit