Alfa Aesar™ Borane-trimethylamine complex, 97%

Manufacturer: Alfa Aesar™ 4296409

Quantity 10g

 View more versions of this product

Catalog No. AA4296409

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-22-9
    Molecular Formula (CH3)3N·BH3
    Formula Weight 72.95
    MDL Number MFCD00012420
    UN Number UN1325
    Beilstein 3905060
    Chemical Name or Material Borane-trimethylamine complex

    Specifications

    Formula Weight 72.95
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 75-22-9
    Boiling Point 172°C
    UN Number UN1325
    Melting Point 93° to 95°C
    Beilstein 3905060
    Sensitivity Moisture Sensitive
    Density 0.81g/mL
    Health Hazard 1 Danger
    MDL Number MFCD00012420
    Quantity 10g
    Flash Point 64°C (147°F)
    Physical Form Powder
    Molecular Formula (CH3)3N·BH3
    Health Hazard 2 GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    TSCA TSCA
    RTECS Number PA0525000
    Chemical Name or Material Borane-trimethylamine complex
    Assay 97%

    GHS H Statement
    H228-H315-H319-H335
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit