Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Borate, 0.5M buffer soln., pH 8.5, low endotoxin

Click to view available options
Quantity:
100mL
250mL
1330-43-4
MFCD00163147
1330-43-4
MFCD00163147
Concentration 0.5M buffer soln.
Physical Form Liquid
pH 8.5
Color Colorless
Quantity 100mL
Chemical Name or Material Borate, low endotoxin