Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Borate, 0.5M buffer soln., pH 9.0, low endotoxin

Click to view available options
Quantity:
100mL
250mL
1330-43-4
MFCD00163147
1330-43-4
MFCD00163147
Concentration 0.5M buffer soln.
Physical Form Liquid
pH 9.0
Color Colorless
Quantity 100mL
Chemical Name or Material Borate, low endotoxin

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA