Borate Buffer, pH 9.5, for Ammonia and Organic Nitrogen Analysis, Ricca Chemical

For Analysis of Ammonia and organic Nitrogen

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 10401

Packaging Poly Natural
Quantity 4L

 View more versions of this product

Catalog No. 1040-1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,1303-96-4,1310-73-2
    CAS Min % 97.0,0.46,0.07
    CAS Max % 100.0,0.48,0.07
    Physical Form Liquid
    Chemical Name or Material Borate Buffer, pH 9.5

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 4L
    CAS Min % 97.0,0.07,0.46
    CAS Max % 0.48,0.07,100.0
    CAS 7732-18-5,1303-96-4,1310-73-2
    Packaging Poly Natural
    Chemical Name or Material Borate Buffer, pH 9.5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit