Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bordeaux R, pure, Thermo Scientific™

Catalog Number AC198050250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
5858-33-3
C20H12N2Na2O7S2
502.423
MFCD00003887
MRVNLKITQIBNKU-LSHDGIFOSA-L
Acid Red 17, C.I. 16180, Fast Red B, 3-Hydroxy-4-(1-naphthylazo)-2, 7-naphthalenedisulfonic acid, disodium salt
9570215
disodium;(4Z)-4-(naphthalen-1-ylhydrazinylidene)-3-oxonaphthalene-2,7-disulfonate
C1=CC=C2C(=C1)C=CC=C2NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-].[Na+].[Na+]
5858-33-3
C20H12N2Na2O7S2
502.423
MFCD00003887
MRVNLKITQIBNKU-LSHDGIFOSA-L
Acid Red 17, C.I. 16180, Fast Red B, 3-Hydroxy-4-(1-naphthylazo)-2, 7-naphthalenedisulfonic acid, disodium salt
9570215
disodium;(4Z)-4-(naphthalen-1-ylhydrazinylidene)-3-oxonaphthalene-2,7-disulfonate
C1=CC=C2C(=C1)C=CC=C2NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-].[Na+].[Na+]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Beilstein 16, I, 305
Infrared Spectrum Authentic
Formula Weight 502.41
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Bordeaux to Brown/Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Bordeaux R
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit