Learn More
BORG5 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302382A

Description
The recommended shelf life for this product is 1 year from date of receipt.
CDC42 is a member of the Rho GTPase family that regulates multiple cellular activities, including actin polymerization. The protein encoded by this gene is a CDC42 binding protein that mediates actin cytoskeleton reorganization at the plasma membrane. This protein is secreted and is primarily found in bone marrow.Specifications
BORG5 | |
Polyclonal | |
Unconjugated | |
CDC42EP1 | |
BORG5, CEP1, MSE55, 55 kDa bone marrow stromal/endothelial cell protein, binder of Rho GTPases 5, cdc42 effector protein 1, serum constituent protein, serum protein MSE55 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11135 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q00587 | |
CDC42EP1 | |
Between 300 and 350 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.