Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, 2% (w/v) Solution, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
10043-35-3
10043-35-3
Quantity 4 L
Grade Chemical Solution
Packaging Poly Bottle