Boric Acid, 4% (w/v) with Bromocresol Green-Methyl Red, for Kjeldahl Analysis, Ricca Chemical

For Analysis of Nitrogen (Kjeldahl)

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 10701

Packaging Poly Bottle
Quantity 4L

 View more versions of this product

Catalog No. 1070-1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,10043-35-3, 62625-32-5, 845-10-3,10043-35-3,62625-32-5,845-10-3
    CAS Min % 94.0,3.77,9.0E-4,7.0E-4
    CAS Max % 98.0,3.92,9.0E-4,7.0E-4
    Physical Form Liquid
    Chemical Name or Material Boric Acid, With Bromocresol Green-Methyl Red Indicator

    Specifications

    Physical Form Liquid
    Color Clear reddish-orange
    Quantity 4L
    CAS 7732-18-5,10043-35-3, 62625-32-5, 845-10-3,845-10-3,10043-35-3,62625-32-5
    Concentration or Composition Notes 4% (w/v)
    Solvent or Matrix Aqueous Solution
    Packaging Poly Bottle
    CAS Max % 9.0E-4,7.0E-4,98.0,3.92
    CAS Min % 7.0E-4,94.0,3.77,9.0E-4
    Chemical Name or Material Boric Acid, With Bromocresol Green-Methyl Red Indicator

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit