Boric acid, 99.8%, Alfa Aesar

Manufacturer: Alfa Aesar 12680A7

 View more versions of this product

Catalog No. AA12680A7

  Check Availability
Add to Cart

    Specifications

    Formula Weight 61.83
    Quantity 5kg
    Color White
    EINECS Number 233-139-2
    Physical Form Granular
    Chemical Name or Material Boric acid
    Assay Percent Range 99.8%
    Molecular Formula BH3O3
    MDL Number MFCD00011337
    Synonym Boracic acid
    Health Hazard 1 H360-H303
    Storage Note 1 Hygroscopic
    Appearance White granular
    Odor Odorless
    CAS 10043-35-3

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit