Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, ACS Reagent, Reagents

Catalog No. 01338186
Click to view available options
Quantity:
500 g
3 kg
10 kg
50 kg

Boric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym Orthoboric Acid, Boracic Acid, Borofax, Trihydroxyborane
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Colorless/White
Melting Point 168°C to 170°C
Boiling Point 300°C
Packaging Poly Bottle
Specific Gravity 1.435
Quantity 3 kg
Physical Form Solid
Grade ACS
Chemical Name or Material Boric Acid, ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.