Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Crystal, ACS, 99.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10043-35-3
10043-35-3
Quantity 500 g
CAS Max % 1
Calcium (Ca) 5e-05
Chloride 1e-05
Percent Purity 0.995
Grade ACS
Heavy Metals 1e-05
Insoluble in Methanol 5e-05
Iron (Fe) 1e-05
Packaging Poly Bottle
Phosphate 1e-05
Sulfate 0.0001
Show More Show Less

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.