Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Crystal, NF, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10043-35-3
BH3O3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
OB(O)O
10043-35-3
BH3O3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
OB(O)O

BO112, 10043-35-3, H3BO3

Spectrum™ Boric Acid, Crystal, NF is used as an antimicrobial preservative, a buffering agent and as a tonicity adjusting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities..
Quantity 50 kg
CAS Max % 1
Percent Purity 99.5 to 100.5%
Loss on Drying 0.005
Packaging Fiber Drum