Boric Acid (Crystalline/Certified ACS), Fisher Chemical

Packaging Fiber Drum
Quantity 325 lb.

 View more versions of this product

Catalog No. A73-325LB

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 10043-35-3
    Physical Form Powder/Solid
    Assay Percent Range ≥99.5 %
    Molecular Formula H3BO3
    Formula Weight 61.83
    MDL Number MFCD00011337
    Synonym Borofax
    Chemical Name or Material Boric Acid
    Assay ≥99.5%
    Grade Certified ACS Grade

    Specifications

    Melting Point 169°C
    Physical Form Powder/Solid
    Formula Weight 61.83
    Quantity 325 lb.
    Color White
    pH 3.8 to 4.8
    Vapor Pressure 2.7mbar at 20°C
    Health Hazard 3 Emergency Overview
    May impair fertility. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Ingestion may cause gastrointestinal irritation, nausea, vomiting and diarrhea. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:2
    Flammability:0
    Instability:1
    Sulfate 0.01% max.
    Assay Percent Range ≥99.5 %
    Calcium (Ca) 0.005% max.
    Chloride 0.001% max.
    Synonym Borofax
    Heavy Metals (as Pb) 0.001% max.
    Insoluble in Methanol 0.005% max.
    Iron (Fe) 0.001% max.
    Non_volatile Matter 0.05% max.
    MDL Number MFCD00011337
    CAS 10043-35-3
    ChemAlert Storage Symbol Gray
    Additional Information Vapor Pressure: 2.7mbar at 20°C
    Packaging Fiber Drum
    Molecular Formula H3BO3
    Health Hazard 2 WARNING!
    Identification Pass Test
    Phosphate 0.001% max.
    Chemical Name or Material Boric Acid
    Assay ≥99.5%
    Grade Certified ACS Grade
     

    WARNING!

    Emergency Overview
    May impair fertility. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause central nervous system effects. Ingestion may cause gastrointestinal irritation, nausea, vomiting and diarrhea. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:2
    Flammability:0
    Instability:1

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit