Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
1kg
Packaging:
Poly Bottle

Chemical Identifiers

10043-35-3
BH3O3
61.831
KGBXLFKZBHKPEV-UHFFFAOYSA-N
7628
CHEBI:33118
boric acid
B(O)(O)O

High quality chemicals for laboratory and specialized industrial use

Quantity 1kg
Formula Weight 61.83
Physical Form Granular
Packaging Poly Bottle
Specific Gravity 1L = 1.43kg
Chemical Name or Material Boric Acid
Grade Baker Analyzed™ ACS Reagent