Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boric Acid, Granular, NF, EP, BP, JP, 99.5-100.5%, Spectrum™ Chemical
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Boric Acid, Granular, NF, EP, BP, JP is used as an antimicrobial preservative, a buffering agent and as a tonicity adjusting agent. Spectrum Chemical NF, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
pH | 3.8-4.8 |
CAS Max % | 100% |
Packaging | Poly Pail |
Quantity | 2.5 kg |
Percent Purity | 99.5 to 100.5% |
Grade | Multi-Compendial |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction