Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Boric Acid, Molecular biology grade, OmniPur™, Calbiochem™,

Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Drum, fiber
Plastic Bottle
Plastic Drum
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O
10043-35-3
BH3O3
61.83
MFCD00011337
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe
7628
CHEBI:33118
boric acid
OB(O)O

Recommended Storage

15°C to 30°C

Color White
Packaging Plastic Bottle
Quantity 1 kg
Physical Form Granular Powder

Recommended Storage : 15°C to 30°C