Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Powder 99+%, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
10043-35-3
BH3O3
61.831
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe
7628
CHEBI:33118
boric acid
B(O)(O)O
10043-35-3
BH3O3
61.831
KGBXLFKZBHKPEV-UHFFFAOYSA-N
orthoboric acid,boracic acid,borofax,boron hydroxide,boron trihydroxide,three elephant,basilit b,trihydroxyborone,h3bo3,flea prufe
7628
CHEBI:33118
boric acid
B(O)(O)O
Lewis acid
Quantity 2.5kg
Color White
Additional Information Particle size: retained on 250 micron sieve: <3%
Assay Percent Range 99+%
Linear Formula H3BO3
Fieser 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70
Merck Index 15, 1339
Chloride 10ppm max.
Chlorine (Cl) 10ppm max.
Formula Weight 61.83
Physical Form Coarse Crystals
Percent Purity ≥99%
Grade Extra Pure
Iron (Fe) 10ppm max.
Odor Odorless
Packaging Plastic bottle
Solubility Solubility in water: 49.5g/L (20°C). Other solubilities: 1g/18mL cold alcohol - 1g/6mL boiling alcohol, 1g/4mL glycerol, nacl, licl, mineral acids decrease the solubility, kno3,k2so4,kcl,nano3,na2so4 increase the sol.
Sulfate 145ppm max.
Melting Point 169.0°C
Chemical Name or Material Boric acid
Show More Show Less

GHS P Statement May damage fertility. May damage the unborn child.

Danger