Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Powder, BiotechGrade, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
10043-35-3
BH3O3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
OB(O)O
10043-35-3
BH3O3
61.83
KGBXLFKZBHKPEV-UHFFFAOYSA-N
boric acid
OB(O)O

B1141, 10043-35-3, H3BO3

Spectrum™ Boric Acid, Powder, BiotechGrade is a weak acid of boron often used as an antiseptic, insecticide, flame retardant, neutron absorber, or precursor to other chemical compounds. It exists in the form of colorless crystals or a white powder that dissolves in water. Boric acid is poisonous if taken internally or inhaled in large quantities.
Quantity 2.5 kg
CAS Max % 100%
Arsenic (As) 8 ppm
Percent Purity 99.5 to 100.5%
DNase RNase Protease act None detected
Heavy Metals 0.002%
Loss on Drying 0.5%
Packaging Poly Pail
pH Range 3.8-4.8
Sulfate 0.01%