Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Powder, Technical, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10043-35-3
10043-35-3
Quantity 45 kg
CAS Max % 1
Grade Technical
Packaging Fiber Drum

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.