Boric Anhydride (Laboratory), Fisher Chemical

Packaging Glass Bottle
Quantity 500g

 View more versions of this product

Catalog No. A76-500

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 1303-86-2
    Physical Form Solid
    Assay Percent Range ≥98 %
    Molecular Formula B2O3
    Formula Weight 69.62
    MDL Number MFCD00011315
    Synonym Boron Trioxide
    Chemical Name or Material Boric Anhydride
    Assay ≥98%
    Grade Laboratory Grade

    Specifications

    Formula Weight 69.62
    Boiling Point 1860°C
    Melting Point 450°C
    Quantity 500g
    Viscosity [NULL]
    Physical Form Solid
    Color White
    pH 4
    Density 2.4
    Health Hazard 3 Emergency Overview
    May impair fertility. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

    NFPA

    Health:2
    Flammability:0
    Instability:1
    Identification Pass Test
    Packaging Glass Bottle
    MDL Number MFCD00011315
    CAS 1303-86-2
    Assay Percent Range ≥98 %
    Molecular Formula B2O3
    ChemAlert Storage Symbol Gray
    Synonym Boron Trioxide
    Health Hazard 2 WARNING!
    Chemical Name or Material Boric Anhydride
    Assay ≥98%
    Grade Laboratory Grade
     

    WARNING!

    Emergency Overview
    May impair fertility. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

    NFPA

    Health:2
    Flammability:0
    Instability:1

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit