Alfa Aesar™ Boron, AAS solution standard, Specpure™, B 1000µg/mL

Manufacturer: Alfa Aesar™ 88055AE

Quantity 100mL

 View more versions of this product

Catalog No. AA88055AE

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula H3BO3 in H2O
    MDL Number MFCD00134034
    Chemical Name or Material Boron, AAS standard solution, B 1000μg/mL
    Grade Specpure™

    Specifications

    Physical Form Liquid
    TSCA TSCA
    MDL Number MFCD00134034
    Quantity 100mL
    Molecular Formula H3BO3 in H2O
    Chemical Name or Material Boron, AAS standard solution, B 1000μg/mL
    Grade Specpure™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit