Alfa Aesar™ Boron nitride, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 40461KJ

Quantity 5 pieces

 View more versions of this product

Catalog No. AA40461KJ

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10043-11-5
    Formula Weight 24.82
    MDL Number MFCD00011317
    Merck Index 14,1346
    Chemical Name or Material Boron nitride, Hot pressed rod, 9.5mm dia x 305mm long

    Specifications

    Physical Form Hot Pressed Rod
    Formula Weight 24.82
    Odor Odorless
    CAS 10043-11-5
    TSCA TSCA
    Merck Index 14,1346
    Quantity 5 pieces
    MDL Number MFCD00011317
    Chemical Name or Material Boron nitride, Hot pressed rod, 9.5mm dia x 305mm long
    Assay 99.5%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit