Boron nitride, 99.5% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1107818

Quantity 50g

 View more versions of this product

Catalog No. AA1107818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10043-11-5
    Physical Form -325 Mesh Powder
    Assay Percent Range 99.5% (metals basis)
    Molecular Formula BN (hexagonal)
    Formula Weight 24.82
    MDL Number MFCD00011317
    Chemical Name or Material Boron nitride

    Specifications

    Formula Weight 24.82
    Quantity 50g
    EINECS Number 233-136-6
    Physical Form -325 Mesh Powder
    Assay Percent Range 99.5% (metals basis)
    MDL Number MFCD00011317
    CAS 10043-11-5
    Molecular Formula BN (hexagonal)
    Chemical Name or Material Boron nitride

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit