Learn More
Boron nitride, Aerosol Refractory Paint, 97+%, (balance B{2}O{3}), Thermo Scientific Chemicals
Description
Boron nitride is used to make alloys, semiconductors, nuclear reactors and lubricants. It is also used in the fabrication and luminescence study of monolayered boron nitride quantum dots. Further, it acts as a starting material to form boron nitride nanotubes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Quantity | 13 oz. |
UN Number | UN1950 |
Solubility Information | Insoluble in water. |
Percent Purity | ≥97% |
Odor | Alcohol-like |
Packaging | Aerosol Spray Can |
Assay | balance B2O3 |
Chemical Name or Material | Boron nitride, Aerosol Refractory Paint |
Safety and Handling
Hazard Category | H222-H319-H335-H336-H350-X |
Hazard Statement | GHS H Statement H220-H319-H336 Extremely flammable gas. Causes serious eye irritation. May cause drowsiness or dizziness. |
Precautionary Statement | P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c |
DOTInformation | Transport Hazard Class: 2.1; Packaging Group: ; Proper Shipping Name: AEROSOLS |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
For Manufacturing and Laboratory Use. Not for beverage use
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.