Alfa Aesar™ Boron nitride, Aerosol Refractory Paint, ≥97%, (balance B2O3)

Manufacturer: Alfa Aesar™ 403890CL

Quantity 5 x 13 oz.

 View more versions of this product

Catalog No. AA403890CL

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10043-11-5
    MDL Number MFCD00011317
    UN Number UN1950
    Chemical Name or Material Boron nitride, Aerosol Refractory Paint

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Packaging Aerosol Spray Can
    Health Hazard 2 GHS H Statement
    H220-H319-H336
    Extremely flammable gas.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    MDL Number MFCD00011317
    CAS 10043-11-5
    UN Number UN1950
    Color White
    DOT Information Transport Hazard Class: 2.1; Packaging Group: ; Proper Shipping Name: AEROSOLS
    TSCA TSCA
    Odor Alcohol-like
    Quantity 5 x 13 oz.
    Health Hazard 1 Danger
    Chemical Name or Material Boron nitride, Aerosol Refractory Paint
    Assay ≥97%, balance B2O3

    GHS H Statement
    H220-H319-H336
    Extremely flammable gas.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit