Learn More
Boron nitride, Refractory Brushable Paint, BN 31%, Thermo Scientific Chemicals
Description
Boron nitride is used as a lubricant and an additive to cosmetic products. It is also used in parts of high-temperature equipment, nanotechnology, polymorph. It is also used in used in xerographic process and laser printers. It is also used in fiberglass, ceramics, wood preservative, nuclear application and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Paste |
Quantity | 1 kg |
Solubility Information | Soluble in water (slightly soluble) at 20°C,and water (soluble) at 95°C. |
IUPAC Name | boranimine |
Chemical Name or Material | Boron nitride |
Safety and Handling
Hazard Category | H301-H332: Toxic if swallowed. Harmful if inhaled. |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.