Alfa Aesar™ Boron nitride, Refractory Brushable Paint, BN 10%

Manufacturer: Alfa Aesar™ 43278A7

Quantity 5kg

 View more versions of this product

Catalog No. AA43278A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10043-11-5
    Molecular Formula BN
    MDL Number MFCD00011317
    Chemical Name or Material Boron nitride, Refractory Brushable Paint

    Specifications

    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Odor Odorless
    Quantity 5kg
    Health Hazard 1 Warning
    Concentration BN 10%
    Physical Form Paste
    MDL Number MFCD00011317
    CAS 10043-11-5
    Color White
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    TSCA TSCA
    Molecular Formula BN
    Chemical Name or Material Boron nitride, Refractory Brushable Paint

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit