Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Boron nitride sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.99% (metals basis)

Magnetron sputtering source

Manufacturer:  Alfa Aesar™ 41052KS

Quantity 1 each

 View more versions of this product

Catalog No. AA41052KS

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

24.82
MFCD00011317
Boron nitride sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick

Specifications

24.82
Odorless
Boron nitride sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MFCD00011317
1 each
99.99%, (metals basis)

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit