Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boron nitride sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.99% (metals basis)
Click to view available options
Quantity:
1 ea.
Specifications
MDL Number | MFCD00011317 |
Formula Weight | 24.82 |
Percent Purity | 99.99% |
Odor | Odorless |
Quantity | 1 ea. |
Chemical Name or Material | Boron nitride sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick |
Assay | (metals basis) |
Safety and Handling
P261-P271-P304+P340-P312-P501c
H335
EINECSNumber : 233-136-6
TSCA : Yes
Recommended Storage : Ambient temperatures