Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron oxide, 98%, -40 mesh, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Fibre drum
Glass Bottle
Plastic bottle
1303-86-2
B2O3
69.61
MFCD00011315
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g
518682
CHEBI:30163
oxo(oxoboranyloxy)borane
B(=O)OB=O
1303-86-2
B2O3
69.61
MFCD00011315
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g
518682
CHEBI:30163
oxo(oxoboranyloxy)borane
B(=O)OB=O
Additional Information total halides impurities (typically): < 0.01%
Assay Percent Range 98%
Linear Formula B2O3
Fieser 01,673; 02,32; 03,33; 09,1349
Merck Index 15,1348
Solubility Information Solubility in water: 36g/L (25°C). Other solubilities: soluble in hot water,soluble in alcohol,glycerol and acids
Formula Weight 69.61
Physical Form Powder
Percent Purity 98%
Density 2.4600g/mL
Packaging Plastic bottle
Solubility Solubility in water: 36g/L (25°C). Other solubilities: soluble in hot water, soluble in alcohol, glycerol and acids
Specific Gravity 2.46
Total Trace Metal Impurities 0.2 % max.
Boiling Point 1860.0°C
Color White
Melting Point 450.0°C
Quantity 5kg
Chemical Name or Material Boron oxide
Show More Show Less

GHS H Statement:
May damage fertility. May damage the unborn child.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 215-125-8