Learn More
Boron oxide, 98%, Thermo Scientific Chemicals
Description
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.
Solubility
Soluble in water. Slightly soluble in methanol.
Notes
Hygroscopic and moisture sensitive. Incompatible with acids and strong oxidizing agents.
Specifications
Boiling Point | 1860°C |
Melting Point | ∽450°C |
Quantity | 500 g |
Merck Index | 14,1337 |
Solubility Information | Soluble in water. Slightly soluble in methanol. |
Formula Weight | 69.62 |
Percent Purity | 99% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 2.46 +0.01 |
Show More |
Safety and Handling
Hazard Category | H360FD |
Hazard Statement | GHS H Statement H360 May damage fertility or the unborn child. |
Precautionary Statement | P201-P202-P281-P308+P313-P501c |
EINECSNumber | 215-125-8 |
RTECSNumber | ED7900000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.