Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Boron oxide, Puratronic™, 99.999% (metals basis)

Click to view available options
Quantity:
100g
500g
1303-86-2
B2O3
69.617
MFCD00011315
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g
518682
CHEBI:30163
oxo(oxoboranyloxy)borane
B(=O)OB=O
1303-86-2
B2O3
69.617
MFCD00011315
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g
518682
CHEBI:30163
oxo(oxoboranyloxy)borane
B(=O)OB=O
  • In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
Merck Index 14,1337
Formula Weight 69.62
Physical Form Fused Lump,Approx. 1.0in dia x 0.75in thick
Percent Purity 99.999%
Grade Puratronic™
Density 2.46 g/mL (±)0.01
Odor Odorless
Sensitivity Hygroscopic
Boiling Point 1860°C
Color White
Melting Point ~450°C
Quantity 100g
Chemical Name or Material Boron oxide, Fused Lump, Approx. 1.0 in. dia. x 0.75 in. thick
Assay (metals basis)
Show More Show Less

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360FD

EINECSNumber : 215-125-8

RTECSNumber : ED7900000

TSCA : Yes

Recommended Storage : Ambient temperatures