Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Boron oxide, Puratronic™, (H2O 200ppm), 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 11160QH

Quantity 10 x 55g/cylinder

 View more versions of this product

Catalog No. AA11160QH


  Check Availability
Add to cart

Specifications

Description

  • In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
Description & Specifications

Chemical Identifiers

1303-86-2
MFCD00011315
Boron oxide, H2O 200ppm, Cylinder, 5.0cm dia.
69.62
14,1337
Puratronic™

Specifications

1303-86-2
69.62
14,1337
Odorless
1860°C
≈450°C
55g/cylinder
99.999%, (metals basis)
Cylinder
MFCD00011315
2.46g/mL (±)0.01
Hygroscopic
White
10 x 55g/cylinder
Boron oxide, H2O 200ppm, Cylinder, 5.0cm dia.
Puratronic™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : ED7900000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit