Learn More
Alfa Aesar™ Boron, plasma standard solution, Specpure™, B 10,000µg/mL
Manufacturer:  Alfa Aesar™ 14370AE
Quantity 100mL
 View more versions of this product
Catalog No. AA14370AE
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD00134034
Boron plasma standard solution, B 10000μg/mL
Specpure™
Specifications
Liquid
MFCD00134034
Boron plasma standard solution, B 10000μg/mL
H3BO3 in 1% NH4OH
100mL
Specpure™
Safety and Handling
SDS & Safety Documents
TSCA : TSCA