Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron powder (96+ atomic % B-10), crystalline, -60+100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number AA4425606
Click to view available options
Quantity:
5 g
25 g
7440-42-8
B
10.81
MFCD00134034 MFCD00151272
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
5462311
CHEBI:27560
boron
[B]
7440-42-8
B
10.81
MFCD00134034 MFCD00151272
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
5462311
CHEBI:27560
boron
[B]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
UN Number UN3178
Percent Purity 99.9%
Mesh Size −60+100 Mesh
Odor Odorless
Form Crystalline
Assay (metals basis), (≥96 atomic % B-10)
Chemical Name or Material Boron powder
Hazard Category Warning
Hazard Statement GHS H Statement
H228-H302-H332-H335
Flammable solid.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 231-151-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit