Alfa Aesar™ Boron powder (96+ atomic % B-11), crystalline, -100 mesh, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4425714

Quantity 25g

 View more versions of this product

Catalog No. AA4425714

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-42-8
    MDL Number MFCD00134034
    UN Number UN3178
    Chemical Name or Material Boron powder

    Specifications

    Physical Form Crystalline Powder
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P304+P340-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 7440-42-8
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H228-H302-H332-H335
    Flammable solid.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause respiratory irritation.
    Odor Odorless
    TSCA TSCA
    UN Number UN3178
    Mesh Size −100 Mesh
    MDL Number MFCD00134034
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
    Chemical Name or Material Boron powder
    Assay 99.9%, (metals basis), (≥96 atomic % B-11)

    GHS H Statement
    H228-H302-H332-H335
    Flammable solid.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P304+P340-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit