Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boron powder, amorphous, -325 mesh, 90%, Mg nominal 5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 100 g |
UN Number | UN3178 |
Percent Purity | 90% |
Concentration or Composition (by Analyte or Components) | Mg nomin.al 5% |
Mesh Size | −325 Mesh |
Odor | Odorless |
Form | Powder |
Chemical Name or Material | Boron powder, Amorphous |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H228-H302-H332-H335 Flammable solid. Harmful if swallowed. Harmful if inhaled. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement P210-P261-P280-P304+P340-P405-P501a Keep away from heat/sparks/open flames/hot surfaces. No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S. |
EINECSNumber | 231-151-2 |
RTECSNumber | ED7350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction