Alfa Aesar™ Boron powder, amorphous, -325 mesh, 90%, Mg nominal 5%

Manufacturer: Alfa Aesar™ 1133806

Quantity 5g

 View more versions of this product

Catalog No. AA1133806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-42-8
    MDL Number MFCD00134034
    UN Number UN3178
    Chemical Name or Material Boron powder, Amorphous

    Specifications

    Quantity 5g
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P304+P340-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    MDL Number MFCD00134034
    UN Number UN3178
    Health Hazard 2 GHS H Statement
    H228-H302-H332-H335
    Flammable solid.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
    TSCA TSCA
    CAS 7440-42-8
    Mesh Size −325 Mesh
    Physical Form Powder
    Health Hazard 1 Warning
    Concentration Mg nomin.al 5%
    Chemical Name or Material Boron powder, Amorphous
    Assay 90%

    GHS H Statement
    H228-H302-H332-H335
    Flammable solid.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P304+P340-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit