Boron Powder, amorphous and crystalline, -325 mesh, 99% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1133703

Quantity 1g

 View more versions of this product

Catalog No. AA1133703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-42-8
    Physical Form Powder, Amorphous and Crystalline
    Assay Percent Range 99% (Metals basis)
    MDL Number MFCD00134034
    Chemical Name or Material Boron

    Specifications

    Physical Form Powder, Amorphous and Crystalline
    Quantity 1g
    Color Brown
    DOT Information DOT Class: 4.1, Packaging Group: III
    EINECS Number 231-151-2
    Ignition Point 700°C (1292°F)
    Odor Odorless
    Assay Percent Range 99% (Metals basis)
    Appearance Brown powder
    MDL Number MFCD00134034
    CAS 7440-42-8
    Health Hazard 1 H228-H302-H332-H335; Flammable solid. Harmful if swallowed. Harmful if inhaled. May cause respiratory irritation.
    Chemical Name or Material Boron

    H228-H302-H332-H335; Flammable solid. Harmful if swallowed. Harmful if inhaled. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit