Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron standard solution, for AAS, 1mg/mL B in water, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Plastic bottle
7440-42-8
B
10.81
MFCD00134034
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
5462311
CHEBI:27560
boron
[B]
7440-42-8
B
10.81
MFCD00134034
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
5462311
CHEBI:27560
boron
[B]
For atomic adsorption spectroscopy
Solubility Solubility in water: soluble in water.
Concentration 997 to 1003 μg/mL of B in water (20°C)
Merck Index 14, 1343
Formula Weight 10.81
Physical Form Solution
Grade AAS
Packaging Plastic bottle
Color Undesignated
Quantity 500mL
Chemical Name or Material Boron standard solution

EINECSNumber : 231-151-2

RTECSNumber : ED7350000

TSCA : TSCA