Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Boron tribromide, 1M soln. in heptane

Manufacturer:  Alfa Aesar™ H32785AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH32785AE


  Check Availability
Add to cart

Specifications

Description

Applications
It is an excellent demethylating or dealkylating agent for the cleavage of ethers, also with subsequent cyclization, often in the production of pharmaceuticals. The use of boron tribromide for effecting complete demethylation of aryl methyl ethers at, or below, room temperature. Phase-Vanishing Reactions that Use Fluorous Media as a Phase Screen. Facile, Controlled Bromination of Alkenes by Dibromine and Dealkylation of Aromatic Ethers by Boron Tribromide.

Solubility
Reacts with water.

Notes
Moisture & Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, moisture, bases.
Description & Specifications

Chemical Identifiers

10294-33-4
250.52
UN3384
BBr3
MFCD00011312
Boron tribromide

Specifications

10294-33-4
250.52
UN3384
90°C
−4°C (25°F)
BBr3
MFCD00011312
1M soln. in heptane
100mL
Boron tribromide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H300-H330-H304-H314-H318-H336
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal if inhaled.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit