Alfa Aesar™ Boron tribromide, 1M soln. in heptane

Manufacturer: Alfa Aesar™ H32785AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH32785AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10294-33-4
    Molecular Formula BBr3
    Formula Weight 250.52
    MDL Number MFCD00011312
    UN Number UN3384
    Chemical Name or Material Boron tribromide

    Specifications

    Formula Weight 250.52
    TSCA TSCA
    Molecular Formula BBr3
    MDL Number MFCD00011312
    UN Number UN3384
    Boiling Point 90°C
    Quantity 500mL
    Flash Point −4°C (25°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H300-H330-H304-H314-H318-H336
    Highly flammable liquid and vapor.
    Fatal if swallowed.
    Fatal if inhaled.
    May be fatal if swallowed and enters airways.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Concentration 1M soln. in heptane
    CAS 10294-33-4
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
    Chemical Name or Material Boron tribromide

    Description

    Applications
    It is an excellent demethylating or dealkylating agent for the cleavage of ethers, also with subsequent cyclization, often in the production of pharmaceuticals. The use of boron tribromide for effecting complete demethylation of aryl methyl ethers at, or below, room temperature. Phase-Vanishing Reactions that Use Fluorous Media as a Phase Screen. Facile, Controlled Bromination of Alkenes by Dibromine and Dealkylation of Aromatic Ethers by Boron Tribromide.

    Solubility
    Reacts with water.

    Notes
    Moisture & Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, moisture, bases.

    GHS H Statement
    H225-H300-H330-H304-H314-H318-H336
    Highly flammable liquid and vapor.
    Fatal if swallowed.
    Fatal if inhaled.
    May be fatal if swallowed and enters airways.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit