Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron tribromide, 1M solution in methylene chloride, ACROS Organics™

Click to view available options
Quantity:
10mL
100mL
800mL
Packaging:
Glass bottle
10294-33-4,75-09-2
BBr3
250.52
MFCD00011312
ILAHWRKJUDSMFH-UHFFFAOYSA-N
boron tribromide,borane, tribromo,boron bromide,tribromoboron,hsdb 327,tribromoboran,boron tribrornide,boron tri bromide,boron tri-bromide
25134
tribromoborane
B(Br)(Br)Br
10294-33-4,75-09-2
BBr3
250.52
MFCD00011312
ILAHWRKJUDSMFH-UHFFFAOYSA-N
boron tribromide,borane, tribromo,boron bromide,tribromoboron,hsdb 327,tribromoboran,boron tribrornide,boron tri bromide,boron tri-bromide
25134
tribromoborane
B(Br)(Br)Br
C-X bond formation reagent, Lewis acid
Linear Formula BBr3
Fieser 01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43
Merck Index 15, 1349
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol,soluble in alcohol and ccl4
Formula Weight 250.52
Physical Form Solution
Concentration 0.9 to 1.1M
Density 1.4670g/mL
Packaging Glass bottle
Solubility Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in alcohol and ccl4
Specific Gravity 1.467
Storage Information Darkens on storage
Color Yellow to Brown
Quantity 100mL
Chemical Name or Material Boron tribromide, 1M Solution in Methylene Chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Fatal if swallowed.
Suspected of causing cancer.
Reacts violently with water.
May cause drowsiness or dizziness.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 233-657-9