Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Boron tribromide, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 3584118

Quantity 50g

 View more versions of this product

Catalog No. AA3584118


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10294-33-4
MFCD00011312
14,1347
250.52
UN2692
Boron tribromide

Specifications

10294-33-4
250.52
UN2692
2.643g/mL
Moisture Sensitive
Light yellow
50g
99.999%, (metals basis)
Liquid
MFCD00011312
14,1347
Acrid
90°C
−46°C
Boron tribromide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H330-H314-H318
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BORON TRIBROMIDE

RTECSNumber : ED7400000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit