Alfa Aesar™ Boron tribromide, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 3584109

Quantity 10g

 View more versions of this product

Catalog No. AA3584109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10294-33-4
    Formula Weight 250.52
    MDL Number MFCD00011312
    UN Number UN2692
    Merck Index 14,1347
    Chemical Name or Material Boron tribromide

    Specifications

    Formula Weight 250.52
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H300-H330-H314-H318
    Fatal if swallowed.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Odor Acrid
    Physical Form Liquid
    UN Number UN2692
    Melting Point −46°C
    Quantity 10g
    Health Hazard 1 Danger
    RTECS Number ED7400000
    TSCA TSCA
    CAS 10294-33-4
    Merck Index 14,1347
    Boiling Point 90°C
    Color Light yellow
    Density 2.643g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BORON TRIBROMIDE
    Sensitivity Moisture Sensitive
    MDL Number MFCD00011312
    Chemical Name or Material Boron tribromide
    Assay 99.999%, (metals basis)

    GHS H Statement
    H300-H330-H314-H318
    Fatal if swallowed.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit