Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron trichloride, 1M solution in methylene chloride, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
10294-34-5, 75-09-2
BCl3
117.16
MFCD00011313
FAQYAMRNWDIXMY-UHFFFAOYSA-N
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
25135
trichloroborane
B(Cl)(Cl)Cl
10294-34-5, 75-09-2
BCl3
117.16
MFCD00011313
FAQYAMRNWDIXMY-UHFFFAOYSA-N
boron trichloride, borane, trichloro, boron chloride, trichloroboron, borontrichloride, chlorure de bore french, unii-k748471rag, hsdb 326, boron trichloride un1741 poison gas
25135
trichloroborane
B(Cl)(Cl)Cl
Lewis acid
Linear Formula BCl3
Concentration 0.9 to 1.1M
Fieser 01,67; 02,34; 03,31; 04,42; 05,50; 06,65; 09,62; 13,43; 15,44
Density 1.3260g/mL
Merck Index 15, 1350
Formula Weight 117.17
Physical Form Liquid
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: may decompose.
Specific Gravity 1.326
Storage Information May darken on storage
Color Colorless to Brown
Quantity 800mL
Chemical Name or Material Boron trichloride, 1M Solution in Methylene Chloride
Show More Show Less

GHS H Statement
Suspected of causing cancer. Causes severe skin burns and eye damage. Toxic if inhaled. Toxic if swallowed. May cause drowsiness or dizziness. Reacts violently with water.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

EINECSNumber : 233-658-4

RTECSNumber : ED1925000

TSCA : TSCA